siRNA Details
| Gene Symbol: DLEU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AJ412054.1 | - | LIT1886 BCMS |
3 | 546 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

DLEU1| Gene Symbol: DLEU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AJ412054.1 | - | LIT1886 BCMS |
3 | 546 | |