siRNA Details
| Gene Symbol: CARMIL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | Y10518.1 | - | 2 | 156 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AY927483.3 | - | 2 | 172 | Chr. 6 - 25527378 - 25556825 [-] on Build GRCh38 | |

CARMIL1
Genomic Map