siRNA Details
| Gene Symbol: KCNIP4-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY072607.1 | - | 1 | 218 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AY072610.3 | - | 574 | Chr. - - [-] on Build | ||

KCNIP4-IT1
Genomic Map