siRNA Details
| Gene Symbol: LINC01150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120534.1 | 4 | 1128 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | U62668.3 | - | 4 | 1129 | Chr. 11 - 1896759 - 1908359 [+] on Build GRCh38 | |

LINC01150
Genomic Map