siRNA Details
| Gene Symbol: DGCR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_110533.1 | 5 | 777 | |||
| GenBank mRNA | X91348.1 | - | LIT1227 DGCR5 |
5 | 749 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

DGCR5| Gene Symbol: DGCR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_110533.1 | 5 | 777 | |||
| GenBank mRNA | X91348.1 | - | LIT1227 DGCR5 |
5 | 749 | |