siRNA Details
| Gene Symbol: SNRPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF400494.1 | - | LIT2208 IC-SNURF-SNRPN 3 prime noncoding transcripts |
2, 3 | 160 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNRPN| Gene Symbol: SNRPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF400494.1 | - | LIT2208 IC-SNURF-SNRPN 3 prime noncoding transcripts |
2, 3 | 160 | |