siRNA Details
| Gene Symbol: SNRPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF400497.1 | - | LIT2205 IC-SNURF-SNRPN 3 prime noncoding transcripts |
12 | 1694 | |
| AY362866.1 | - | 3 | 249 | |||

SNRPN
Genomic Map