siRNA Details
| Gene Symbol: LINC01152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_110124.1 | 3 | 2016 | |||
| NR_110132.1 | 3 | 1506 | ||||
| GenBank mRNA | D43770.1 | - | LIT1584 CMPD |
3 | 1854 | |

LINC01152
Genomic Map