siRNA Details
| Gene Symbol: LINC01152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_110124.1 | 3 | 1216 | |||
| NR_110132.1 | 3 | 706 | ||||
| GenBank mRNA | D43770.1 | - | LIT1584 CMPD |
3 | 1054 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01152| Gene Symbol: LINC01152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_110124.1 | 3 | 1216 | |||
| NR_110132.1 | 3 | 706 | ||||
| GenBank mRNA | D43770.1 | - | LIT1584 CMPD |
3 | 1054 | |