siRNA Details
| Gene Symbol: SNRPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF400486.1 | - | LIT2209 IC-SNURF-SNRPN 3 prime noncoding transcripts |
11 | 1576 | |
| AL832758.1 | - | 8 | 1057 | |||
| BC080646.1 | - | 8 | 1066 | |||

SNRPN
Genomic Map