siRNA Details
| Gene Symbol: PBDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927529.1 | - | LIT3537 AY927529 |
1 | 357 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PBDC1| Gene Symbol: PBDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY927529.1 | - | LIT3537 AY927529 |
1 | 357 | |