siRNA Details
| Gene Symbol: NPSR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY312367.1 | - | LIT3327 AAA1 |
5 | 601 | |
| U85653.1 | - | 5 | 564 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NPSR1-AS1| Gene Symbol: NPSR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AY312367.1 | - | LIT3327 AAA1 |
5 | 601 | |
| U85653.1 | - | 5 | 564 | |||