siRNA Details
| Gene Symbol: LOC101929106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110052.1 | 5 | 2421 | |||
| GenBank mRNA | AK127178.1 | - | 5 | 2421 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929106| Gene Symbol: LOC101929106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110052.1 | 5 | 2421 | |||
| GenBank mRNA | AK127178.1 | - | 5 | 2421 | ||