siRNA Details
| Gene Symbol: LOC101929106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110052.1 | 1, 2 | 338 | |||
| GenBank mRNA | AK127178.1 | - | 1, 2 | 338 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000356133.3 | - | 2 | 338 | Chr. 3 - 187203548 - 187207514 [-] on Build GRCh38 | |

LOC101929106
Genomic Map