siRNA Details
| Gene Symbol: RAB18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256410.1 | NP_001243339.1 | 8 | 3827 | ||
| NM_001256411.1 | NP_001243340.1 | 6 | 3673 | |||
| NM_001256412.1 | NP_001243341.1 | 5 | 3548 | |||
| NM_001256415.1 | NP_001243344.1 | 6 | 3668 | |||
| NM_021252.4 | NP_067075.1 | 7 | 3740 | |||
| NR_046172.1 | 6 | 3678 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR610854.1 | - | 1 | 448 | Chr. 10 - 27540527 - 27542214 [+] on Build GRCh38 | |

RAB18
Genomic Map