siRNA Details
| Gene Symbol: RAB18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256410.1 | NP_001243339.1 | 8 | 4937 | ||
| NM_001256411.1 | NP_001243340.1 | 6 | 4783 | |||
| NM_001256412.1 | NP_001243341.1 | 5 | 4658 | |||
| NM_001256415.1 | NP_001243344.1 | 6 | 4778 | |||
| NM_021252.4 | NP_067075.1 | 7 | 4850 | |||
| NR_046172.1 | 6 | 4788 | ||||

RAB18
Genomic Map