siRNA Details
| Gene Symbol: CCNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001240.3 | NP_001231.2 | 9 | 3053 | ||
| NM_001277842.1 | NP_001264771.1 | 8 | 2889 | |||
| XM_017020197.1 | XP_016875686.1 | 8 | 2564 | |||
| GenBank mRNA | BX537514.1 | - | 1 | 24 | ||

CCNT1
Genomic Map