siRNA Details
Gene Symbol: LINC00943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038256.1 | 2 | 607 | |||
GenBank mRNA | AK091521.1 | - | 2 | 1080 | ||
AY070435.1 | - | 729 | ||||
BC039096.1 | - | 2 | 589 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038256.1 | 2 | 607 | |||
GenBank mRNA | AK091521.1 | - | 2 | 1080 | ||
AY070435.1 | - | 729 | ||||
BC039096.1 | - | 2 | 589 |