siRNA Details
| Gene Symbol: LINC00943 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038256.1 | 2 | 341 | |||
| GenBank mRNA | AK091521.1 | - | 2 | 814 | ||
| AY070435.1 | - | 463 | ||||
| BC039096.1 | - | 2 | 323 | |||
| DB450910.1 | - | 2 | 371 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AL538253.3 | - | 1 | 195 | Chr. 12 - 126742736 - 126745093 [+] on Build GRCh38 | |

LINC00943
Genomic Map