siRNA Details
| Gene Symbol: LINC00943 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038256.1 | 2 | 869 | |||
| GenBank mRNA | AK091521.1 | - | 2 | 1342 | ||
| AY070435.1 | - | 991 | ||||
| BC039096.1 | - | 2 | 851 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00943| Gene Symbol: LINC00943 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038256.1 | 2 | 869 | |||
| GenBank mRNA | AK091521.1 | - | 2 | 1342 | ||
| AY070435.1 | - | 991 | ||||
| BC039096.1 | - | 2 | 851 | |||