siRNA Details
| Gene Symbol: LOC91548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK026812.1 | - | 6 | 976 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC91548| Gene Symbol: LOC91548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK026812.1 | - | 6 | 976 | ||