siRNA Details
| Gene Symbol: LOC91548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK026812.1 | - | 6 | 1108 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000421933.1 | - | 6 | 1109 | Chr. 1 - 211681764 - 211687459 [+] on Build GRCh38 | |

LOC91548
Genomic Map