siRNA Details
| Gene Symbol: HMBOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005273635.1 | XP_005273692.1 | 11 | 13399 | ||
| XM_017013823.1 | XP_016869312.1 | 11 | 13434 | |||
| XM_017013824.1 | XP_016869313.1 | 11 | 13251 | |||
| GenBank mRNA | BX648950.1 | - | 1 | 4115 | ||

HMBOX1
Genomic Map