siRNA Details
| Gene Symbol: MAP3K13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC026249.1 | - | 1 | 639 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAP3K13| Gene Symbol: MAP3K13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC026249.1 | - | 1 | 639 | ||