siRNA Details
| Gene Symbol: KXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011528272.2 | XP_011526574.1 | 1 | 392 | ||
| GenBank mRNA | AK125365.1 | - | 1 | 525 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KXD1| Gene Symbol: KXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011528272.2 | XP_011526574.1 | 1 | 392 | ||
| GenBank mRNA | AK125365.1 | - | 1 | 525 | ||