siRNA Details
| Gene Symbol: SPATS2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092917.1 | - | 1 | 220 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK097196.1 | - | 1 | 2736 | Chr. 2 - 200474114 - 200477515 [+] on Build GRCh38 | |

SPATS2L
Genomic Map