siRNA Details
| Gene Symbol: RPS15A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK023088.1 | - | 5 | 1680 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK127316.1 | - | 1 | 3314 | Chr. 16 - 18782968 - 18786795 [-] on Build GRCh38 | |

RPS15A
Genomic Map