siRNA Details
| Gene Symbol: RBMS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003792.2 | NP_001003792.1 | 14 | 5586 | ||
| NM_001003793.2 | NP_001003793.1 | 15 | 5640 | |||
| NM_001177711.1 | NP_001171182.1 | 14 | 5592 | |||
| XM_005265063.1 | XP_005265120.1 | 12 | 5297 | |||
| XM_005265065.4 | XP_005265122.1 | 12 | 4990 | |||
| XM_017006179.1 | XP_016861668.1 | 14 | 6041 | |||
| XM_017006180.1 | XP_016861669.1 | 13 | 5993 | |||
| XM_017006181.1 | XP_016861670.1 | 13 | 5990 | |||
| XM_017006182.1 | XP_016861671.1 | 14 | 6069 | |||
| GenBank mRNA | AK093863.1 | - | 1 | 581 | ||

RBMS3
Genomic Map