siRNA Details
| Gene Symbol: C9orf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017015230.1 | XP_016870719.1 | 16 | 4959 | ||
| XM_017015231.1 | XP_016870720.1 | 14 | 4662 | |||
| XM_017015237.1 | XP_016870726.1 | 13 | 4495 | |||
| GenBank mRNA | AF043897.1 | - | 1 | 1694 | ||

C9orf3
Genomic Map