siRNA Details
| Gene Symbol: MCM3AP-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110567.1 | 1 | 47 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF426262.1 | - | 1 | 31 | Chr. 21 - 46229247 - 46243001 [+] on Build GRCh38 | |

MCM3AP-AS1
Genomic Map