siRNA Details
| Gene Symbol: PTPN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005401.4 | NP_005392.2 | 19 | 12990 | ||
| GenBank mRNA | AK056963.1 | - | 1 | 656 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTPN14| Gene Symbol: PTPN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005401.4 | NP_005392.2 | 19 | 12990 | ||
| GenBank mRNA | AK056963.1 | - | 1 | 656 | ||