siRNA Details
| Gene Symbol: GAS2L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303130.1 | NP_001290059.1 | 9 | 2498 | ||
| NM_001303131.1 | NP_001290060.1 | 10 | 2985 | |||
| NM_174942.2 | NP_777602.1 | 10 | 2619 | |||
| GenBank mRNA | BX649059.1 | - | 1 | 1434 | ||

GAS2L3
Genomic Map