siRNA Details
| Gene Symbol: ZC3H14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011537162.2 | XP_011535464.1 | 10 | 4221 | ||
| GenBank mRNA | AL832086.1 | - | 1 | 62 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZC3H14| Gene Symbol: ZC3H14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011537162.2 | XP_011535464.1 | 10 | 4221 | ||
| GenBank mRNA | AL832086.1 | - | 1 | 62 | ||