siRNA Details
| Gene Symbol: ANKRD54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011529878.2 | XP_011528180.1 | 1 | 362 | ||
| XM_017028590.1 | XP_016884079.1 | 1 | 362 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD54| Gene Symbol: ANKRD54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011529878.2 | XP_011528180.1 | 1 | 362 | ||
| XM_017028590.1 | XP_016884079.1 | 1 | 362 | |||