siRNA Details
| Gene Symbol: ERV18-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126787.1 | - | 1 | 1009 | ||
| Gene Symbol: UNC13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017022223.1 | XP_016877712.1 | 1 | 147 | ||

ERV18-1
Genomic Map