siRNA Details
Gene Symbol: LOC101929413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109866.1 | 1 | 118 | |||
GenBank mRNA | BC033557.1 | - | 1 | 118 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG721017.1 | - | 1 | 123 | Chr. 20 - 10894812 - 10909272 [-] on Build GRCh38 |
