siRNA Details
Gene Symbol: LOC101929413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109866.1 | 7 | 1470 | |||
GenBank mRNA | BC033557.1 | - | 7 | 1470 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000448859.1 | - | 7 | 1470 | Chr. 20 - 10875333 - 10909272 [-] on Build GRCh38 |
