siRNA Details
| Gene Symbol: LOC101929413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109866.1 | 7 | 1599 | |||
| GenBank mRNA | BC033557.1 | - | 7 | 1599 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929413| Gene Symbol: LOC101929413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109866.1 | 7 | 1599 | |||
| GenBank mRNA | BC033557.1 | - | 7 | 1599 | ||