siRNA Details
| Gene Symbol: C1orf116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BG150416.1 | - | 1 | 202 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF007835.1 | - | 1 | 319 | Chr. 1 - 207018401 - 207018900 [+] on Build GRCh38 | |

C1orf116
Genomic Map