siRNA Details
| Gene Symbol: LOC101929721 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125988.1 | 3 | 200 | |||
| GenBank mRNA | AK131006.1 | - | 3 | 201 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000420071.1 | - | 3 | 200 | Chr. 1 - 49257411 - 49269285 [+] on Build GRCh38 | |

LOC101929721
Genomic Map