siRNA Details
| Gene Symbol: LOC101929721 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125988.1 | 4 | 429 | |||
| GenBank mRNA | AK131006.1 | - | 4 | 430 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929721| Gene Symbol: LOC101929721 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125988.1 | 4 | 429 | |||
| GenBank mRNA | AK131006.1 | - | 4 | 430 | ||