siRNA Details
| Gene Symbol: ST6GALNAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX648274.1 | - | 2 | 5634 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC041381.1 | - | 1 | 808 | Chr. 1 - 76633426 - 76637546 [+] on Build GRCh38 | |

ST6GALNAC3
Genomic Map