siRNA Details
| Gene Symbol: SPAG5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL157421.1 | - | 1 | 590 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | HY044173.1 | - | 3 | 337 | Chr. 17 - 28599133 - 28616399 [+] on Build GRCh38 | |

SPAG5-AS1
Genomic Map