siRNA Details
| Gene Symbol: LOC101929109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134278.1 | 2 | 1523 | |||
| GenBank mRNA | BC043525.1 | - | 2 | 1523 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929109| Gene Symbol: LOC101929109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134278.1 | 2 | 1523 | |||
| GenBank mRNA | BC043525.1 | - | 2 | 1523 | ||