siRNA Details
Gene Symbol: LOC101927363 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134672.1 | 2 | 205 | |||
GenBank mRNA | BC043520.1 | - | 2 | 205 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI831373.1 | - | 2 | 206 | Chr. 4 - 32351038 - 32353211 [+] on Build GRCh38 |
