siRNA Details
| Gene Symbol: LOC101927363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134672.1 | 2 | 526 | |||
| GenBank mRNA | BC043520.1 | - | 2 | 526 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI831373.1 | - | 2 | 526 | Chr. 4 - 32351038 - 32353211 [+] on Build GRCh38 | |

LOC101927363
Genomic Map