siRNA Details
| Gene Symbol: OK/SW-CL.36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB064670.1 | - | 1 | 3408 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OK/SW-CL.36| Gene Symbol: OK/SW-CL.36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB064670.1 | - | 1 | 3408 | ||