siRNA Details
| Gene Symbol: LINC00837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038374.1 | 3 | 2796 | |||
| NR_038375.1 | 2 | 2694 | ||||
| NR_038376.1 | 2 | 2573 | ||||
| GenBank mRNA | AL080132.1 | - | 1 | 1516 | ||
| BC028422.1 | - | 2 | 2618 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00837| Gene Symbol: LINC00837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038374.1 | 3 | 2796 | |||
| NR_038375.1 | 2 | 2694 | ||||
| NR_038376.1 | 2 | 2573 | ||||
| GenBank mRNA | AL080132.1 | - | 1 | 1516 | ||
| BC028422.1 | - | 2 | 2618 | |||