siRNA Details
Gene Symbol: C9orf173-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038389.1 | 4 | 1152 | |||
GenBank mRNA | AK097764.1 | - | 4 | 1152 |
Gene Symbol: FAM166A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006717108.2 | XP_006717171.2 | 1 | 426 | ||
XM_011518694.1 | XP_011516996.1 | 1 | 426 | |||
XM_011518695.1 | XP_011516997.1 | 1 | 426 | |||
XM_011518696.2 | XP_011516998.1 | 1 | 426 | |||
XM_011518697.1 | XP_011516999.1 | 1 | 426 | |||
XM_017014718.1 | XP_016870207.1 | 1 | 426 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000612170.1 | - | 1152 | Chr. - - [-] on Build |
