siRNA Details
| Gene Symbol: C9orf173-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038389.1 | 1 | 257 | |||
| GenBank mRNA | AK097764.1 | - | 1 | 257 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9orf173-AS1| Gene Symbol: C9orf173-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038389.1 | 1 | 257 | |||
| GenBank mRNA | AK097764.1 | - | 1 | 257 | ||